Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Hyaluronidase-1 (from cells)
Human recombinant Hyaluronidase-1 (from cells)
Catalog # PRSI91-778
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Hyaluronidase-1 (from cells)
Catalog # PRSI91-778
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Hyaluronidase-1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    47.18 kD
  • Sequence:
    Phe22-Trp435
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Hyaluronidase-1 (HYAL1) is a secreted lysosomal hyaluronidase that belongs to the glycosyl hydrolase 56 family. HYAL1 contains one EGF-like domain and is highly expressed in the liver, kidney, and heart, but it is weakly expressed in the lung, placenta, and skeletal muscle. HYAL1 is thought to be involved in cell proliferation, migration, and differentiation. It may play a role in promoting tumor progression and blocking the TGFB1-enhanced cell growth. Mutations in HYAL1 are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.