Human recombinant Dipeptidase 1 (from cells)
Catalog # PRSI91-648
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:Dipeptidase 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:42.1 kD
- Sequence:Asp17-Ser385
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Dipeptidase 1 (DPEP1) is a kidney membrane enzyme that belongs to the peptidase M19 family. DPEP1 is a homodimer and is inhibited by L-penicillamine. DPEP1 hydrolyses a variety of dipeptides and is implicated in renal metabolism of glutathione and its conjugates. DPEP1 is responsible for hydrolysis of the beta-lactam ring of antibiotics, such as penem and carbapenem. DPEP1 may play an important role in the regulation of leukotriene activity. DPEP1 expression in cancer is significantly higher than that in normal tissue. However, DPEP1 expression decreased with pathological differentiation, lymph-node and distant metastasis.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.