Order Entry
Denmark
ContactUsLinkComponent
 
Mouse recombinant carboxypeptidase M
  PRSI91-622
 :  
Mouse recombinant carboxypeptidase M
  PRSI91-622
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    Carboxypeptidase M
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    47.5 kD
  • Sequence:
    Leu18-Ser423
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of PBS,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Carboxypeptidase M (CPM) belongs to the peptidase M14 family, and exists in cell membrane. The protein binds 1 zinc ion per subunit, and cleavage of C-terminal arginine or lysine residues from polypeptides. CPM specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localised degradation of extracellular proteins.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.