Order Entry
Denmark
ContactUsLinkComponent
 
Human recombinant Hydroxyacid Oxidase 1 (from E. coli)
Catalog # PRSI91-259
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Hydroxyacid Oxidase 1 (from E. coli)
Catalog # PRSI91-259
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-Trx, 6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Hydroxyacid Oxidase 1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    58.6 kD
  • Sequence:
    Met1-Ile370
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Hydroxyacid Oxidase 1 (HAO1) is an enzyme that belongs to the FMN-Dependent alpha-Hydroxy Acid Dehydrogenase family. HAO1 contains 1 FMN Hydroxy Ccid Dehydrogenase domain. HAO1 is expressed primarily in the liver and pancreas. This protein has 2-Hydroxyacid Oxidase activity. Most HAO1 is active on the 2-Carbon substrate Glycolate, but it can also be active on 2-Hydroxy fatty acids, with higher activity towards 2-Hydroxy Palmitate and 2-Hydroxy Octanoate.

  • Fusion-Tag: N-Trx, 6 His tag

Fusion-Tag: N-Trx, 6 His tag

This recombinant protein can be used for biological assays. For research use only.