Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Galactose Mutarotase (from E. coli)
Human recombinant Galactose Mutarotase (from E. coli)
Catalog # PRSI91-258
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Galactose Mutarotase (from E. coli)
Catalog # PRSI91-258
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Galactose Mutarotase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    38.8 kD
  • Sequence:
    Ala2-Ala342
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Galactose Mutarotase (GALM) is a cytoplasmic enzyme that belongs to the Aldose Epimerase family. GALM is a Mutarotase that converts alpha-Aldose to the beta -Anomer. GALM is active on D-Glucose, L-Arabinose, D-Xylose, D-Galactose, Maltose and Lactose. GALM may be required for normal Galactose metabolism by maintaining the equilibrium of alpha- and beta - anomers of Galactose.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.