Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Malate dehydrogenase (MDH)
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:37.5 kD
- Sequence:Ser2-Ala334
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Malate Dehydrogenase, Cytoplasmic (MDH1) is an enzyme which belongs to the MDH Type 2 sub-family of LDH/MDH superfamily. MDH1 is involved in the Citric Acid Cycle that catalyses the conversion of Malate into Oxaloacetate (using NAD+) and vice versa. MDH1 should not be confused with Malic Enzyme, which catalyses the conversion of Malate to Pyruvate, producing NADPH. MDH1 also participates in Gluconeogenesis, the synthesis of Glucose from smaller molecules. Pyruvate in the mitochondria is acted upon by Pyruvate Carboxylase to form Pxaloacetate, a Citric Acid Cycle intermediate. In order to transport the Oxaloacetate out of the Mitochondria, Malate Dehydrogenase reduces it to Malate, and it then traverses the inner Mitochondrial membrane. Once in the cytosol, the Malate is oxidised back to Oxaloacetate by MDH1. Finally, Phosphoenol-Pyruvate Carboxy Kinase (PEPCK) converts Oxaloacetate to Phosphoenol Pyruvate.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.