Order Entry
Denmark
ContactUsLinkComponent
 
Human recombinant Carbonic Anhydrase 8 (from E. coli)
  PRSI91-095
 :  
Human recombinant Carbonic Anhydrase 8 (from E. coli)
  PRSI91-095
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Carbonic Anhydrase 8
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    34.04 kD
  • Sequence:
    Ala2-Gln290
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 500 mM NaCl, 1 mM DTT, pH 8.5 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Carbonic Anhydrase 8 (CA8) belongs to the alpha-carbonic anhydrase family. Alpha-carbonic anhydrase is a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Because CA8 has some sequence similarity with other known carbonic anhydrase genes, it was firstly called as CA-related protein. Nevertheless, CA8 does not have a carbonic anhydrase catalytic activity. Defects in CA8 are the cause of cerebellar ataxia mental retardation and dysequilibrium syndrome type 3 (CMARQ3), which is a congenital cerebellar ataxia associated with dysarthia, quadrupedal gait and mild mental retardation.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.