Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Ph20 hyaluronidase (from HEK293 cells)
Human recombinant Ph20 hyaluronidase (from HEK293 cells)
Catalog # PRSI96-626
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant Ph20 hyaluronidase (from HEK293 cells)
Catalog # PRSI96-626
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Enzyme type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Enzyme name:
    Ph20 Hyaluronidase
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular weight:
    52.1 kDa
  • Formulation:
    Lyophilized, 50 mM Tris, 100 mM NaCl, pH7.0 - 7.4
  • Tested applications:
    Western Blot

Specifications

About this item