Order Entry
Denmark
ContactUsLinkComponent
Human recombinant IL-2 R gamma (from cells)
Human recombinant IL-2 R gamma (from cells)
  PRSI92-451
 :  
Human recombinant IL-2 R gamma (from cells)
  PRSI92-451
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CD132|Cytokine receptor common subunit gamma|IL2RG|Interleukin-2 receptor subunit gamma|IL-2R subunit gamma|gammaC|IL-2 receptor subunit gamma
  • Protein/peptide name:
    IL-2 R gamma
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    55.4 kD
  • Sequence:
    Leu23-Ala262
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

 

 

IL2RG contains one fibronectin type-III domain. IL2RG is an important signalling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. IL2RG interacts with SHB upon interleukin stimulation and HTLV-1 accessory protein p12I. Defects in IL2RG are the cause of X-linked combined immunodeficiency (XCID) and severe combined immunodeficiency X-linked T-cell-negative /B-cell-positive / NK-cell-negative (XSCID).

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.