Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:ITIH3|SHAP|ITI heavy chain H3|Inter-alpha-inhibitor heavy chain 3|Serum-derived hyaluronan-associated protein|Inter-alpha-trypsin inhibitor heavy chain H3|ITI-HC3
- Protein/peptide name:ITI heavy chain H3
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:70.4 kD
- Sequence:Leu35-Asp651
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
ITIH3, which is short for Inter-alpha-trypsin inhibitor heavy chain H3, is a 890 aa. protein. It is secreted expression, and belongs to the ITIH family. I-alpha-I plasma protease inhibitors are assembled from one or two heavy chains (H1, H2 or H3) and one light chain, bikunin. Inter-alpha-inhibitor (I-alpha-I) is composed of H1, H2 and bikunin, inter-alpha-like inhibitor (I-alpha-LI) of H2 and bikunin, and pre-alpha-inhibitor (P-alpha-I) of H3 and bikunin. ITTH3 may act as a carrier of hyaluronan in serum or as a binding protein between hyaluronan and other matrix protein, including those on cell surfaces in tissues to regulate the localisation, synthesis and degradation of hyaluronan which are essential to cells undergoing biological processes.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.