Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Resistin (from E. coli)
Human recombinant Resistin (from E. coli)
Catalog # PRSI92-180
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Resistin (from E. coli)
Catalog # PRSI92-180
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Cysteine-rich secreted protein FIZZ3|Adipose tissue-specific secretory factor|Cysteine-rich secreted protein A12-alpha-like 2|HXCP1|Resistin|RETN|FIZZ3|RSTN|C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein|ADSF
  • Protein/peptide name:
    Resistin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    10.6 kD
  • Sequence:
    Lys19-Pro108
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Acetic acid,pH 3.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Resistin known as adipose tissue-specific secretory factor (ADSF) or C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein (XCP1) that seems to suppress insulin ability to stimulate glucose uptake into adipose cells. The length of the resistin pre-peptide in human is 108 amino acid residues and in the mouse and rat it is 114 aa; the molecular weight is ~12.5 kDa. Resistin is a cytokine whose physiologic role has been the subject of much controversy regarding its involvement with obesity and type II diabetes mellitus (T2DM). Resistin has been shown to cause "high levels of 'bad' cholesterol (low-density lipoprotein or LDL), increasing the risk of heart disease, resistin increases the production of LDL in human liver cells and also degrades LDL receptors in the liver. Potentially links obesity to diabetes.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.