Human recombinant valosin-containing (from E. coli)
Catalog # PRSI92-101
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:TER ATPase|VCP|Valosin-Containing Protein|Transitional Endoplasmic Reticulum ATPase|15S Mg(2+)-ATPase p97 Subunit
- Protein/peptide name:Valosin-Containing
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:66.4 kD
- Sequence:Ala2-Asn589
- Formulation:Supplied as a 0.2 um filtered solution of 50mM TrisHCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Valosin-Containing Protein (VCP) is a nuclear protein that belongs to the AAA ATPase family. VCP is a putative ATP-binding protein involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. It is necessary for the fragmentation of Golgi stacks during mitosis and their reassembly after mitosis. VCP has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation. VCP participates in the formation of the transitional endoplasmic reticulum (tER) and regulates E3 ubiquitin-protein ligase activity of RNF19A.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag