Human recombinant ZBED1 (from E. coli)
PRSI92-021
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:dREF Homolog|Putative Ac-like Transposable Element|ALTE|Zinc Finger BED Domain-Containing Protein 1|KIAA0785|DREF|ZBED1|TRAMP
- Protein/peptide name:ZBED1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:12.5 kD
- Sequence:Asn3-Glu100
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Zinc Finger BED Domain-Containing Protein 1 (ZBED1) contains one BED-type zinc finger and is found in the cell nucleus. ZBED1 is widely expressed, highly in heart, skeletal muscle, spleen and placenta. The expression of ZBED1 is usually linked to the cell cycle. During the G1/S phase, the expression is increasing. During the S/G2 phage, the expression reaches to the highest, and then decreasing. ZBED1 exists in homodimer forms, which can bind to 5’-TGTCCG[CT]GA[CT]A-3’ DNA elements, that can be found in the promoter regions of a number of gene related to cell proliferation.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag