Order Entry
Denmark
ContactUsLinkComponent
Human recombinant neuronal cell adhesion molecule (from cells)
Human recombinant neuronal cell adhesion molecule (from cells)
Catalog # PRSI91-816
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant neuronal cell adhesion molecule (from cells)
Catalog # PRSI91-816
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    KIAA0343|Nr-CAM|Neuronal surface protein Bravo|Ng-CAM-related|hBravo|gCAM-related cell adhesion molecule|Neuronal cell adhesion molecule|NRCAM
  • Protein/peptide name:
    Neuronal cell adhesion molecule
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    91.2 kD
  • Sequence:
    Gln25-Asn600
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Neuronal cell adhesion molecule(NRCAM) is a single-pass type I membrane protein ,containing 5 fibronectin type-III domains and 6 Ig-like C2-type (immunoglobulin-like) domains.It belongs to the immunoglobulin superfamily. NrCAM is engaged in such biological processes as axonal fasciculation, cell-cell adhesion, central nervous system development, clustering of voltage-gated sodium channels, neuron migration, positive regulation of neuron differentiation, regulation of axon extension, and synaptogenesis. It also may play a role in the molecular assembly of the nodes of Ranvier. NrCAM effects are also linked with different recognition processes and signal transduction pathways regulating cell differentiation, proliferation, or migration

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-Fc tag