Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Apolipoprotein C-II (from E. coli)
Human recombinant Apolipoprotein C-II (from E. coli)
Catalog # PRSI91-800
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Apolipoprotein C-II (from E. coli)
Catalog # PRSI91-800
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    APC2 and APOC2|ApoC2|Apolipoprotein C-II|Apolipoprotein C2
  • Protein/peptide name:
    Apolipoprotein C-II
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    10 kD
  • Sequence:
    Met1-Glu82
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mMPB,150mM NaCL,30%glycerol,pH7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

APOC2 activates the lipoprotein lipase in capillaries, which hydrolyzes triglycerides and thus provides free fatty acids for cells. APOC2 is component of the very low density lipoprotein (VLDL) fraction in plasma. It is also an activator of several triacylglycerol lipases. The association of APOC2 with plasma chylomicrons, VLDL, and HDL is reversible, a function of the secretion and catabolism of triglyceride-rich lipoproteins, and changes rapidly. Defects in APOC2 are the cause of hyperlipoproteinemia type 1B (HLPP1B) which characterized by hypertriglyceridemia, xanthomas, and increased risk of pancreatitis and early atherosclerosis.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag