Order Entry
Denmark
ContactUsLinkComponent
Human recombinant uroplakin 2 (from cells)
Human recombinant uroplakin 2 (from cells)
Catalog # PRSI91-723
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant uroplakin 2 (from cells)
Catalog # PRSI91-723
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    UP2|UPII|Uroplakin II|UPK2|Uroplakin-2
  • Protein/peptide name:
    Uroplakin 2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    14.84 kD
  • Sequence:
    Asp26-Gly155
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Uroplakin-2 is a single-pass type I membrane protein that belongs to the uroplakin-2 family. Uroplakin-2 is a component of the asymmetric unit membrane (AUM) and expressed in the ureter, a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. Uroplakin-2 forms heterodimer with UPK1A that is necessary for exiting out of the endoplasmic reticulum (ER). Uroplakin-2 may play an important role in regulating the assembly of the AUM. AUM is believed to strengthen the urothelium by preventing cell rupture during bladder distention.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag