Human recombinant high mobility Group B3 (from cells)
PRSI91-678
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:High Mobility Group Protein B3|HMG4|HMGB3|HMG-2a|High Mobility Group Protein 2a|HMG-4|High Mobility Group Protein 4|HMG2A
- Protein/peptide name:High Mobility Group B3
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:24 kD
- Sequence:Met1-Glu200
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
High Mobility Group Protein B3 (HMGB3) belongs to the HMGB family. Members of the HMG box subfamily are thought to be have an important role in DNA replication, nucleosome assembly and transcription. HMGB3 binds preferentiallly single-stranded DNA and unwinds double stranded DNA. HMGB3 consists of 200 amino acids and is localized to the cell nucleus. It contains two HMG box DNA-binding domain. HMGB3 binds preferentially single-stranded DNA and unwinds double stranded DNA.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag