Human recombinant VISTA (from cells)
Catalog # PRSI91-633
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-Fc tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:B7-H5|C10orf54|Platelet receptor Gi24|SISP1|Stress-induced secreted protein-1|Sisp-1|PD-1H
- Protein/peptide name:VISTA
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:45.3 kD
- Sequence:Phe33-Ala194
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
platelet receptor Gi24 is a single-pass type I membrane protein, and located at the cell surface. The protein can be cleaved by MMP14, and stimulate MMP14-mediated MMP2 activation. It is participated in the BMP signaling pathway. It also regulates the CD4-pasitive, alpha-beta T cell proliferation, and T cell cytokine production negatively. However, the protein can regulate stem cell differentiation positively.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag