Order Entry
Denmark
ContactUsLinkComponent
Human recombinant GFER (from E. coli)
Human recombinant GFER (from E. coli)
Catalog # PRSI91-585
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant GFER (from E. coli)
Catalog # PRSI91-585
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    hERV1|HPO|GFER|FAD-linked sulfhydryl oxidase ALR|Hepatopoietin|ALR|Augmenter of liver regeneration
  • Protein/peptide name:
    GFER
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.3 kD
  • Sequence:
    Met1-Asp125
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

GFER is a hepatotrophic growth factor and flavin-linked sulfhydryl oxidase which belongs to the Erv1/ALR family of proteins. GFER is widely expressed in various human tissues. They are two isoforms of this protein. Isoform 1 could regenerate the redox-active disulfide bonds in CHCHD4/MIA40, a chaperone essential for disulfide bond formation and protein folding in the mitochondrial intermembrane space. The reduced form of CHCHD4/MIA40 forms a transient intermolecular disulfide bridge with GFER/ERV1, resulting in regeneration of the essential disulfide bonds in CHCHD4/MIA40, while GFER/ERV1 becomes re-oxidized by donating electrons to cytochrome c or molecular oxygen. Isoform 2 may act as an autocrine hepatotrophic growth factor promoting liver regeneration. GFER could also induce the expression of S-adenosylmethionine decarboxyl-ase and ornithine decarboxylases (ODC). S-adenosylmethionine decarboxyl-ase and ornithine decarboxylases play an important role in the synthesis of polyamines.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag