Order Entry
Denmark
ContactUsLinkComponent
Human recombinant TF (from cells)
Human recombinant TF (from cells)
Katalog # PRSI91-434
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant TF (from cells)
Katalog # PRSI91-434
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    F3|Coagulation Factor III|Thromboplastin|CD142|Tissue Factor|TF
  • Protein/peptide name:
    TF
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    25.76 kD
  • Sequence:
    Gly34-Glu251
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Tissue Factor (TF) is a single-pass type I membrane glycoprotein member of the tissue factor family. TF expression is highly dependent upon cell type. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII. TF initiates blood coagulation by forming a complex with circulating factor VII or VIIa. The complex activates factors IX or X by specific limited protolysis. TF plays a role in normal hemostasis by initiating the cell-surface assembly and propagation of the coagulation protease cascade.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag