Order Entry
Denmark
ContactUsLinkComponent
Human recombinant ERMAP (from cells)
Human recombinant ERMAP (from cells)
Catalog # PRSI91-422
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ERMAP (from cells)
Catalog # PRSI91-422
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    ERMAP|hERMAP|Erythroid Membrane-Associated Protein|SC|Radin Blood Group Antigen|RD|Scianna Blood Group Antigen
  • Protein/peptide name:
    ERMAP
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    14.79 kD
  • Sequence:
    His30-Ala155
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Human Erythroid Membrane-Associated Protein (ERMAP) is a cell surface transmembrane protein that belongs to the immunoglobulin superfamily. It is hghly expressed in bone marrow and to a lower extent in leukocytes, thymus, lymph node and spleen. ERMAP contains 1 B30.2/SPRY domain and 1 Ig-like V-type (immunoglobulin-like) domain. It may serve as an erythroid cell receptor, possibly as a mediator of cell adhesion. ERMAP is responsible for the Scianna/Radin blood group system. Two transcript variants encoding the same protein have been found for this gene ERMAP.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag