Human recombinant Apolipoprotein H (from cells)
Katalog # PRSI91-388
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:APC inhibitor|Beta-2-Glycoprotein I|Beta-2-Glycoprotein 1|B2GPIBeta(2)GPI|APOH|B2G1|Activated Protein C-Binding Protein|Apolipoprotein H|Anticardiolipin Cofactor|Apo-H
- Protein/peptide name:Apolipoprotein H
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:37.29 kD
- Sequence:Gly20-Ser345
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Apolipoprotein H (ApoH) is a 50 kDa variably glycosylated member of the complement control superfamily of proteins. Human ApoH is a major phospholipid binding protein and an important component to measure in the assessment of anti-phospholipid syndrome. Hepatocyte-derived ApoH binds to negatively charged phospholipids . It circulates as a component of lipoprotein particles and as a lipid-free serum protein. Human ApoH is also more specific than anti-cardiolipin antibodies and its presence correlates better with thrombotic risk. Mature human ApoH shares 76% and 82% aa sequence identity with mouse and rat ApoH.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag