Order Entry
Denmark
ContactUsLinkComponent
Human recombinant beta-Defensin 1 (from E. coli)
Human recombinant beta-Defensin 1 (from E. coli)
Catalog # PRSI91-108
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant beta-Defensin 1 (from E. coli)
Catalog # PRSI91-108
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    HBD1|BD1|Defensin Beta 1|DEFB1|Beta-Defensin 1|hBD-1|BD-1
  • Protein/peptide name:
    beta-Defensin 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    5.07 kD
  • Sequence:
    Gly22-Lys68
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 130mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays

Specifications

About this item

beta -Defensin 1 (DEFB1) is a member of the beta -defensin family, which is highly expressed by epithelial cells. beta -defensins are expressed as the C-terminal portion of precursors and are released by proteolytic cleavage of a signal peptide. beta -defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds. beta -defensin 1 is an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. Defects in beta -Defensin-1 contribute to asthma diagnosis, with apparent gender-specific effects in human. beta -defensin 1 may also play a role in the pathogenesis of severe sepsis. In addition, beta -defensin 1 is associated with induction profiles in gingival keratinocytes.

  • This recombinant protein can be used for biological assays. For research use only.