Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:IL-TIF Alpha|IL-TIF|IL-22|IL-22a|IL-10-Related T-Cell-Derived-Inducible Factor|Il22a|Iltif|Interleukin-22a|Interleukin-22|Il22|Iltifa
- Protein/peptide name:IL22
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:16.7 kD
- Sequence:Leu34-Val179
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:For Biological assays
Specifications
About this item
Interleukin-22 (IL-22) was initially identified as a gene induced by IL-9 in mouse T cells and mast cells. Mouse IL-22 cDNA encodes a 179 amino acid residue protein with a putative 33 amino acid signal peptide that is cleaved to generate a 147 amino acid mature protein that shares approximately 79% and 22% sequence identity with human IL22 and IL10, respectively. IL22 has been shown to activate STAT-1 and STAT-3 in several hepatoma cell lines and up-regulate the production of acute phase proteins. IL-22 is produced by normal mouse T cells upon Con A activation. Mouse IL-22 expression is also induced in various organs upon lipopolysaccharide injection, suggesting that IL-22 may be involved in inflammatory responses. The functional IL-22 receptor complex consists of two receptor subunits, IL-22R (previously an orphan receptor named CRF2-9) and IL-10R beta (previously known as CRF2-4), belonging to the class II cytokine receptor family.
- This recombinant protein can be used for biological assays. For research use only.