Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant CD1261 (from HEK293 cells)
Human Recombinant CD1261 (from HEK293 cells)
Catalog # PRSI96-471
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human Recombinant CD1261 (from HEK293 cells)
Catalog # PRSI96-471
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its binding ability in a functional ELISA.Immobilized Human IL-6 R alpha, His Tag at 5μg/ml (100 μL/well),can bind Biotinylated Human IL-6 with a linear range of 4-250 ng/mL. Measured by its ability to enhance the IL6 activity on M1 mouse myeloid leukemia cells.The ED50 for this effect is typically 15-60 ng/mL.
  • Protein synonyms:
    IL-6RA|IL6RA|IL-6R-alpha|IL6RQ|IL-6R-1|IL6R|CD126|gp80
  • Protein/peptide name:
    CD1261
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular weight:
    40 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

Interleukin 6 receptor (IL6R) is also known as CD126 (Cluster of Differentiation 126) , is a potent pleiotropic cytokine that regulates cell growth and differentiation of various tissues, and is known particularly for its role in the immune response and acute phase reactions. IL6R is a protein complex consisting of a IL-6 receptor subunit (IL6R) and interleukin 6 signal transducer Glycoprotein 130. IL6R also denotes the human gene encoding this subunit. Alternatively spliced transcript variants encoding distinct isoforms have been reported. IL6R subunit also shared by many other cytokines. The soluble form of IL6R arises from proteolytic cleavage of membrane-bound IL6Rα, and acts agonistically by making the IL6 ligand accessible to the signal transducer gp130. Dysregulated production of IL6 and IL6R are implicated in the pathogenesis of several inflammatory diseases and malignancies such as multiple myeloma, rheumatoid arthritis, or osteoporosis, and it has been reported that a humanised anti-IL6R monoclonal antibody is a promising agent applicable to the therapeutic approach for IL6 driven diseases. Interleukin-6 receptor has been shown to interact with Interleukin 6 and Ciliary neurotrophic factor.