Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant CD25 (from HEK293 cells)
Human Recombinant CD25 (from HEK293 cells)
Catalog # PRSI96-459
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human Recombinant CD25 (from HEK293 cells)
Catalog # PRSI96-459
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Protein synonyms:
    IL2-RA|p55|IL-2-RA|CD25|IL2RA
  • Protein/peptide name:
    CD25
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    23.6 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

Interleukin-2 receptor subunit alpha (IL2RA) is also known as IL-2R subunit alpha, IL-2-RA, IL2-RA, TAC antigen, p55, CD antigen CD25, is a type I transmembrane glycoprotein. IL2RA is expressed on activated T cells and regulatory T cells, and is capable of binding IL2 with low affinity by itself. However, a ligand-induced high affinity heterotrimeric receptor complex is produced when IL2RA is associated non-covelently with the IL2 receptor beta and gamma chain, and subsequently initiates the intacellular signal pathways such as MAPK or JAK/STAT. On dendritic cells (DC), CD25 has been previously regarded as an activation marker, while both murine and human DC can express CD25, they do not express the beta-chain of the IL-2 receptor, which is indispensable for the execution of IL-2 signaling.