Human recombinant Hsp 90 alpha (from E. coli), GST tag
PRSI96-395
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Tag sequence:GST Tag
- Protein synonyms:HSPCA|Hsp89|EL52|HSP86|HSP 86|HSP90AA1|HSPCAL1|HSPN|Hsp90|HSPC1|HSP90N|HSPCAL4|LAP2|HSP89A|HSP90A
- Protein/peptide name:Hsp 90 alpha
- Purity:>85% as determined by SDS-PAGE.
- Molecular weight:49.3 kDa
- Formulation:Lyophilized, 50 mM HEPES, pH7.0, with 150 mM NaCl
- Tested applications:Western Blot
Heat shock protein HSP 90-alpha (HSP90AA1 or HSP90A) is also known as Heat shock 86 kDa (HSP 86 or HSP86), Renal carcinoma antigen NY-REN-38, HSPC1, HSPCA, EL52, HSP89A, HSP90N, HSPCAL1, HSPCAL4, HSPN, Hsp89, Hsp90, LAP2, which belongs to the heat shock protein 90 family. HSP90AA1 undergoes a functional cycle that is linked to its ATPase activity. This cycle probably induces conformational changes in the client proteins, thereby causing their activation. HSP90AA1 interacts dynamically with various co-chaperones that modulate its substrate recognition, ATPase cycle and chaperone function.