Human recombinant CD69 (from HEK293 cells)
Catalog # PRSI96-143
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:EA1|MLR-3|AIM|GP32/28|CLEC2C|CD69|BL-AC/P26
- Protein/peptide name:CD69
- Purity:>92% as determined by SDS-PAGE.
- Molecular weight:16.8 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot
Specifications
About this item
Early activation antigen CD69 is also known as C-type lectin domain family 2 member C (CLEC2C), Activation inducer molecule (AIM), Early T-cell activation antigen p60, Leukocyte surface antigen Leu-23. CD69 contains one C-type lectin domain. CD69 / CLEC2C is expressed on the surface of activated T-cells, B-cells, natural killer cells, neutrophils, eosinophils, epidermal Langerhans cells and platelets. CD69 is induced by antigens, mitogens or activators of PKC on the surface of T and B-lymphocytes and by interaction of IL-2 with the p75 IL-2R on the surface of NK cells. CD69 / CLEC2C involved in lymphocyte proliferation and functions as a signal transmitting receptor in lymphocytes, natural killer (NK) cells, and platelets.