Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human recombinant BDNF (from <i>E. coli</i>)

Human recombinant BDNF (from E. coli)

10 µG
Catalog # PRSI40-514
Supplier:   ProSci Inc.
Human recombinant BDNF (from <i>E. coli</i>)
Catalog # PRSI40-514
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant BDNF (from E. coli)

10 µG

CAS Number:  
Price
2.720,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

BDNF is a member of the NGF family of neurotrophic growth factors. Like other members of this family, BDNF supports neuron proliferation and survival. BDNF can bind to a low affinity cell surface receptor called LNGFR, which also binds other neurotrophins such as NGF, NT-3 and NT-4. However, BDNF mediates its neurotrophic properties by signaling through a high affinity cell surface receptor called gp145/trkB. BDNF is expressed as the C-terminal portion of a 247 amino acid polypeptide precursor, which also contains a signal sequence of 18 amino acid residue and a propeptide of 110 amino acid residues. Recombinant human BDNF is a 27.0 kDa homodimer of two 119 amino acid subunits linked by strong non-covalent interactions. Human and Mouse BDNF sequences are identical.

  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Documents

Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    PRSI40-514
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    BDNF
  • Formulation:
    Lyophilized
  • Protein synonyms:
    Brain-derived neurotrophic factor|ANON2|BDNF|BULN2|Abrineurin
  • Purity:
    >98%
  • Species:
    Human
  • Source:
    E. coli
  • Biological activity:
    Biological activity was determined by its ability to stimulate the proliferation of rat C6 cells. The expected ED50 for this effect is 0.5-1.0 μg/mL.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.