Order Entry
Denmark
ContactUsLinkComponent
Human recombinant LXN (from E. coli)
Human recombinant LXN (from E. coli)
Catalog # PRSIXW-RP3139
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant LXN (from E. coli)
Catalog # PRSIXW-RP3139
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein synonyms:
    Endogenous carboxypeptidase inhibitor|ECI|TCI|Latexin
  • Protein/peptide name:
    LXN
  • Purity:
    >95%
  • Molecular weight:
    25.5 kDa (Calculated)
  • Tested applications:
    ELISA, Western Blot, MS

Specifications

About this item

Strongly similar to rat latexin (TCI, Rn.11404); may be a tissue carboxypeptidase inhibitor.

  • This recombinant protein can be used for WB, ELISA, MS.

10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.