Specifications
- Antigen name:Protein Phosphatase 1, Regulatory Subunit 1B
- Antigen symbol:PPP1R1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Xenopus,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Neuronal phosphoprotein DARPP-32|IPPD|Dopamine- and cAMP-regulated neuronal phosphoprotein
- Immunogen:DARPP-32 (Thr34) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acids residues surrounding the phospho Thr34 of DARPP-32.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
DARPP-32, a dopamine (DA) and cAMP-regulated ~32k phosphoprotein that is associated with dopaminoceptive neurons bearing D-1 receptors in the basal ganglia. The protein inhibits protein phosphatase I when it is phosphorylated on Thr34. In contrast, when DARPP-32 is phosphorylated on Thr75 the protein acts as an inhibitor of PKA. Phosphorylation of DARPP-32 is thought to play a critical role in the regulation of dopaminergic neurotransmission. In addition, the activity of DARPP-32 is also thought to play important roles in the actions of alcohol, caffeine and Prozac®.
Applications include Dot Blots (DB) and Western Blots (WB). Suitability for Immunohistochemistry (IHC) has not yet been determined. Rabbit anti-DARPP-32 (Thr34) recognizes human, mouse, and rat forms of the protein. When internally tested under ideal conditions the working dilutions were 1:1000 for DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -80°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PPP1R1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus