Specifications
- Antigen name:Interleukin 5
- Antigen symbol:IL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Antigen synonyms:Interleukin-5|EDF|Eosinophil differentiation factor|T-cell replacing factor|TRF|IL-5|B cell differentiation factor I
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIL-5 (human Interleukin-5).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Interleukin 5 (IL-5) is a cytokine produced primarily by activated T lymphocytes. It exists as an antiparallel disulfide linked homodimeric glycoprotein with 115 amino acid residues in each chain. Known also as EDF (eosinophil differentiating factor), functions predominantly as an eosinophilopoietic factor. Analysis of its crystal structure reveals a novel two domain structure, with each domain showing significant homology to the cytokine fold in GMCSF, MCSF, IL-2, IL-4 and growth hormone. Human and mouse IL-5 have 70% amino acid sequence homology.
Anti-hIL-5 specific antibody was purified by affinity chromatography employing immobilized hIL-5 matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: IL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human