Anti-APMAP Mouse Monoclonal Antibody
PRSI38-298
- Antigen name:Adipocyte Plasma Membrane Associated Protein
- Antigen symbol:APMAP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:a|Adipose most abundant gene transcript 1|30 kDa adipocyte complement-related protein|Adipocyte|ACRP30|C1q and collagen domain-containing protein
- Immunogen:Hi-5 Insect cells derived Recombinant Human Adiponectin
- Size:500 µg
- Pk:500 µG
This antibody was purified from cell culture by Protein A affinity chromatography.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type:
Antigen: APMAP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human