Anti-GLI1 Rabbit Polyclonal Antibody
PRSI31-153
- Antigen name:GLI Family Zinc Finger 1
- Antigen symbol:GLI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Zfp5|GLI|Zfp-5|AV235269
- Immunogen:A synthetic peptide corresponding a region of human GLI1.
- Size:100 µg
- Pk:100 µG
GLI1 is a protein which is a member of the Kruppel family of zinc finger proteins. The function of this gene has not been determined; however, it may play a role in normal development gene transcription. Mouse mutation studies indicate possible involvement in human foregut malformation.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GLI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human