Anti-GABRG3 Rabbit Polyclonal Antibody
PRSI31-134
- Antigen name:Gamma-aminobutyric acid (GABA) A Receptor gamma 3
- Antigen symbol:GABRG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Gamma aminobutyric acid GABA A receptor gamma 3|GABRG3|GABAA receptor|GABR G3|Gamma aminobutyric acid receptor subunit gamma 3|Gamma aminobutyric acid A receptor gamma 3|GABRG 3|GABAA receptor subunit gamma 3|GBRG3_HUMAN.
- Immunogen:A synthetic peptide corresponding a region of human GABRG3.
- Size:100 µg
- Pk:100 µG
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GABRG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human