Anti-CHI3L1 Rabbit Polyclonal Antibody
Katalog # PRSI30-782
Supplier: ProSci Inc.
Specifications
- Antigen name:Chitinase 3-like 1
- Antigen symbol:CHI3L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CGP-39|GP-39|YYL-40|GP39|YKL40|HCGP-3P|YKL-40|HC-gp39|hCGP-39|Cartilage glycoprotein-39|ASRT7
- Immunogen:A synthetic peptide corresponding a region of human CHI3L1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
CHI3L1 may play an important role in the capacity of cells to respond to and cope with changes in their environment. CHI3L1 may play a role in tissue remodeling and defense against pathogens. It belongs to the glycosyl hydrolase 18 family. CHI3L1 may play an important role in the capacity of cells to respond to and cope with changes in their environment.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CHI3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: