Order Entry
Denmark
ContactUsLinkComponent
Anti-LZTFL1 Rabbit Polyclonal Antibody
Anti-LZTFL1 Rabbit Polyclonal Antibody
Catalog # PRSI30-403
Supplier:  ProSci Inc.
undefined
Anti-LZTFL1 Rabbit Polyclonal Antibody
Catalog # PRSI30-403
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Leucine Zipper Transcription Factor-like 1
  • Antigen symbol:
    LZTFL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    BBS17
  • Immunogen:
    A synthetic peptide corresponding a region of human LZTFL1.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The function remains unknown.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LZTFL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat