Anti-SLC25A39 Rabbit Polyclonal Antibody
Catalog # PRSI29-929
Supplier: ProSci Inc.
Specifications
- Antigen name:Solute Carrier Family 25 Member A39
- Antigen symbol:SLC25A39
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:CGI-69|CGI69
- Immunogen:A synthetic peptide corresponding a region of human SLC25A39.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC25A39 is a member of the solute carrier family 25 and is known to transport molecules over the mitochondrial membrane.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC25A39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat