Anti-RPF1 Rabbit Polyclonal Antibody
Catalog # PRSI29-471
Supplier: ProSci Inc.
Specifications
- Antigen name:Ribosome Production Factor 1 homolog
- Antigen symbol:RPF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:BXDC5|bA397G5.4
- Immunogen:A synthetic peptide corresponding a region of human BXDC5.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
BXDC5 may be required for ribosome biogenesis.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RPF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat