Anti-CSDC2 Rabbit Polyclonal Antibody
Catalog # PRSI29-420
Supplier: ProSci Inc.
Specifications
- Antigen name:Cold Shock Domain Containing C2
- Antigen symbol:CSDC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:RNA binding protein PIPPin|Csdc2|Cold shock domain containing C2 RNA binding|CSDC2_HUMAN|Cold shock domain-containing protein C2|RNA-binding protein PIPPin.|Cold shock domain containing protein C2|dJ347H13.2|PIPPIN|CSDC 2
- Immunogen:A synthetic peptide corresponding a region of human CSDC2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
CSDC2 is an RNA-binding factor which binds specifically to the very 3'UTR ends of both histone H1 and H3.3 mRNAs, encompassing the polyadenylation signal. It might play a central role in the negative regulation of histone variant synthesis in the developing brain.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CSDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: