Anti-TRPM5 Rabbit Polyclonal Antibody
Catalog # PRSI28-257
Supplier: ProSci Inc.
Specifications
- Antigen name:Transient Receptor Potential Cation Channel Subfamily M member 5
- Antigen symbol:TRPM5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:TRPM5|LTRPC5|Transient receptor potential cation channel subfamily M member 5|MTR1|MLSN1- and TRP-related gene 1 protein
- Immunogen:A synthetic peptide corresponding a region of human TRPM5.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
TRPM5 is a voltage-modulated Ca (2+)-activated, monovalent cation channel (VCAM) that mediates a transient membrane depolarization and plays a central role in taste transduction. It is activated by arachidonic acid in vitro. It may be involved in perception of bitter, sweet and umami tastes. It may also be involved in sensing semiochemicals.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRPM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse