Anti-ZDHHC17 Rabbit Polyclonal Antibody
PRSI26-187
- Antigen name:Zinc Finger, DHHC-type Containing 17
- Antigen symbol:ZDHHC17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:HSPC294|HYPH|HIP14|HIP3
- Immunogen:A synthetic peptide corresponding a region of human ZDHHC17.
- Size:50 µg
- Pk:50 µG
ZDHHC17 is a palmitoyltransferase specific for a subset of neuronal proteins, including SNAP25, DLG4/PSD95, GAD2, SYT1 and HD. It may be involved in the sorting or targeting of critical proteins involved in the initiating events of endocytosis at the plasma membrane. It may be involved in the NF-kappa-B signaling pathway and has transforming activity.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZDHHC17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat