Anti-ADARB2 Rabbit Polyclonal Antibody
Product Details & Documents
ADARB2 is a member of the double-stranded RNA adenosine deaminase family of RNA-editing enzymes and may play a regulatory role in RNA editing. This gene encodes a member of the double-stranded RNA adenosine deaminase family of RNA-editing enzymes and may play a regulatory role in RNA editing. Sequence Note: The RefSeq transcript and protein were derived from genomic sequence to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on alignments.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ADARB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10099-706.pdf
Datasheet
Specifications
- Size:50 µg
- Western blot:Yes
- Cat. No.:PRSI25-654
- Clonality:Polyclonal
- Reactivity:Human
- Form:Lyophilized
- Antigen synonyms:ADARB2
- Immunogen:A synthetic peptide corresponding a region of human ADARB2.
- Host:Rabbit
- Conjugation:Unconjugated
- Antigen symbol:ADARB2
- Antigen name:Adenosine Deaminase, RNA-specific, B2
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



