Specifications
- Antigen name:Zinc Finger CCCH type Containing 7B
- Antigen symbol:ZC3H7B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:RoXaN|Scrg3|Zc2h7b|Gm627|mKIAA1031
- Immunogen:A synthetic peptide corresponding a region of human ZC3H7B.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ZC3H7B is a protein that contains a tetratricopeptide repeat domain. The encoded protein also interacts with the rotavirus non-structural protein NSP3. This gene encodes a protein that contains a tetratricopeptide repeat domain. The encoded protein also interacts with the rotavirus non-structural protein NSP3.
ZC3H7B antibody can be used for detection of ZC3H7B by ELISA at 1:312500. ZC3H7B antibody can be used for detection of ZC3H7B by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZC3H7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat