Anti-SARS SPIKE Rabbit Polyclonal Antibody
Catalog # PRSI3219
Supplier: ProSci Inc.
Specifications
- Antigen name:Sars Spike Protein
- Antigen symbol:SARS SPIKE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:SARS Spike antibody was raised against a synthetic peptide corresponding to amino acids at the amino-terminus of. The sARS Spike glycoprotein.
- Tested applications:E
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
SARS Spike Antibody: A novel coronavirus has recently been identified as the causative agent of SARS (Severe Acute Respiratory Syndrome). Coronaviruses are a major cause of upper respiratory diseases in humans. The genomes of these viruses are positive-stranded RNA approximately 27-31kb in length. SARS infection can be mediated by the binding of the viral spike protein, a glycosylated 139 kDa protein and the major surface antigen of the virus, to the angiotensin-converting enzyme 2 (ACE2) on target cells. This binding can be blocked by a soluble form of ACE2.
Type:
Antigen: SARS SPIKE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus