- Antigen name:Caspase 12
- Antigen symbol:CASP12
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Rabbit
- ImmunoChemistry:Yes
- ImmunFluorescens:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonymer:CASP12P1|CASP-12
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:Caspase-12 antibody was raised against a synthetic peptide corresponding to amino acids within the large cleavage product of murine caspase-12.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Størrelse:100 µg
- Pakn:100 µG
Caspase-12 Antibody: (Large)Three distinct signaling pathways lead to programmed cell death (apoptosis). The death receptor and mitochondrion pathways are the main, in which the key apoptotic proteases capase-8 and caspase-9, respectively, are involved. The endoplasmic reticulum (ER) stress is the third apoptotic pathway and caspase-12 is involved. Caspase-12 is localized to the ER but not to cytoplasm or mitochondrion. Caspase-12 is activated by ER stress, including disruption of ER calcium homeostasis, and mediates ER stress-induced apoptosis. Caspase-12 is co-localized to the ER with several proteins that are involved in Alzheimer's disease including gamma-secretase presenilin and beta-amyloid precursor protein (APP). Caspase-12 mediates cytotoxicity induced by amyloid-beta. Caspase-12 is ubiquitously expressed in mouse tissues.
Type:
Antigen: CASP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat