Order Entry
Denmark
ContactUsLinkComponent
Ferromanganese, high carbon, alloy, NIST-68C, reference standard
Ferromanganese, high carbon, alloy, NIST-68C, reference standard
Catalog # PROONIST-68C
CAS Number:  
Ferromanganese, high carbon, alloy, NIST-68C, reference standard
Catalog # PROONIST-68C
CAS Number:  
Specifications
  • Pk:
    100 g
  • Description:
    Ferromanganese, high carbon
Specifications
About this item
This standard reference material is in the powder form intended for use in checking chemical methods of analysis and in calibration with instrumental methods of analysis.
  • 8 analytes/elements measured
Certifications: Traceable to NIST and SRM.