Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Triclocarban, Reference Standards

Supplier:   USP
Triclocarban, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1681800.
  • 1681815.
  • 1681845.
  • 1681856.
  • Beskrivelse:
  • Triclocarban
  • Triclocarban Related Compound A
  • Triclocarban Related Compound C
  • Triclocarban Mixture Related Compound D
  • Synonym(s):
  • 1,3-Bis(4-chlorophenyl)urea
  • 1,3-Bis(3,4-dichlorophenyl)urea
  • CAS-nr:
  • 101-20-2
  • 1219-99-4
  • 4300-43-0
  • Formel:
  • C₁₃H₉Cl₃N₂O
  • C₁₃H₁₀Cl₂N₂O
  • C₁₃H₈Cl₄N₂O
  • UN No.:
  • 3077
  • 3077
  • 3077

Specifications

Product Family Options

Product Information

  • Beskrivelse
    Standard
    Pakning
    Pakn
    Availability
    Price
  • Triclocarban
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 2.110,00 kr
    2.110,00 kr
    1 * 200 mg
       2.110,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1681800.
    Beskrivelse
    Triclocarban
    Synonym(s)
    CAS-nr
    101-20-2
    Formel
    C₁₃H₉Cl₃N₂O
    UN No.
    3077
  • Triclocarban Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 3.940,00 kr
    3.940,00 kr
    1 * 15 mg
       3.940,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1681815.
    Beskrivelse
    Triclocarban Related Compound A
    Synonym(s)
    1,3-Bis(4-chlorophenyl)urea
    CAS-nr
    1219-99-4
    Formel
    C₁₃H₁₀Cl₂N₂O
    UN No.
    3077
  • Triclocarban Related Compound C
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 3.940,00 kr
    3.940,00 kr
    1 * 15 mg
       3.940,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1681845.
    Beskrivelse
    Triclocarban Related Compound C
    Synonym(s)
    1,3-Bis(3,4-dichlorophenyl)urea
    CAS-nr
    4300-43-0
    Formel
    C₁₃H₈Cl₄N₂O
    UN No.
    3077
  • Triclocarban Mixture Related Compound D
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 3.940,00 kr
    3.940,00 kr
    1 * 15 mg
       3.940,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1681856.
    Beskrivelse
    Triclocarban Mixture Related Compound D
    Synonym(s)
    CAS-nr
    Formel
    UN No.