Syringes, Cavro
Cavro XL syringe with Luer lock termination, 1 ml
Catalog # SGEA008140
Supplier: Trajan Scientific and Medical
Jump to:
Product Details & Documents
These cavro syringes are a type of instrument syringes used in chromatography.
- Luer lock termination
- Gas tight plunger with CAVRO XL mounting hole
Specifications
- Volume:1 ml
- Description:Cavro XL syringe with Luer lock termination
- Seq:01