Order Entry
Denmark
Orders LinkContactUsLinkComponent
Polyethylene Terephthalate, Reference Standards

Polyethylene Terephthalate, Reference Standards

Supplier:   USP
Polyethylene Terephthalate, Reference Standards
Supplier:  USP
Certificates
Polyethylene Terephthalate, Reference Standards

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1546900.
  • 1546922.
  • Description:
  • Polyethylene Terephthalate
  • Polyethylene Terephthalate G
  • Synonym(s):
  • PETG
  • CAS No.:
  • 25038-59-9
  • 25640-14-6
  • Formula:
  • C₁₀H₁₂O₆
  • C₁₈H₂₆O₆

Specifications

Product Family Options

Product Information

  • Standards
    Pk Info
    Pack type
    Pk
    Availability
    Price
  • 1546900.
    Polyethylene Terephthalate
    Polyethylene Terephthalate
    USP reference standard
    3 strips 2'' x 2 1/2'' each
    Strips
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 3.080,00 kr
    3.080,00 kr
    1 * 1 Each
       3.080,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1546900.
    Description
    Polyethylene Terephthalate
    Synonym(s)
    CAS No.
    25038-59-9
    Formula
    C₁₀H₁₂O₆
  • 1546922.
    Polyethylene Terephthalate G
    Polyethylene Terephthalate G
    USP reference standard
    3 Strips
    Strips
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 3.290,00 kr
    3.290,00 kr
    1 * 1 Each
       3.290,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1546922.
    Description
    Polyethylene Terephthalate G
    Synonym(s)
    PETG
    CAS No.
    25640-14-6
    Formula
    C₁₈H₂₆O₆